missing translation for 'onlineSavingsMsg'
Learn More

Crry Antibody (TLD-1C11), Novus Biologicals™

Product Code. 18364004 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364004 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364004 Supplier Novus Biologicals Supplier No. NBP311189

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Crry Monoclonal specifically detects Crry in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Crry
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Functional Assay, Immunoassay
Classification Monoclonal
Clone TLD-1C11
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Complement component (3b/4b) receptor 1-like, Complement component receptor 1-like protein, Complement receptor type-1 related gene Y, Complement regulatory protein Crry, CR, Cr1l, CRY, MCP, mCR, mCRY, p65, Protein p65
Host Species Mouse
Immunogen Rat microglial cells
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12946
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.