missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550940
This item is not returnable.
View return policy
Description
CRP1 Polyclonal Antibody detects endogenous levels of CRP1 protein.
This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation.
Specifications
| CRP1 | |
| Polyclonal | |
| Unconjugated | |
| CSRP1 | |
| AA408841; AA959891; AW545626; CRP; CRP1; CSRP; Csrp 1; Csrp1; CYRP; cysteine and glycine rich protein 1; cysteine and glycine-rich protein 1; cysteine rich protein 1; cysteine-rich protein 1; D1S181E; Epididymis luminal protein 141; epididymis secretory protein Li 286; HEL141; HEL-141; HEL-S-286; LIM-domain protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13007, 1465, 29276 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P21291, P47875, P97315 | |
| CSRP1 | |
| Synthesized peptide derived from the Internal region of human CRP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction