missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Crotonyl-HIST1H2BC (Lys20) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117476
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. The protein has antibacterial and antifungal antimicrobial activity. The main transcript variant of this gene is intronless and encodes a replication-dependent histone that is a member of the histone H2B family. This transcript variant lacks a polyA tail but instead contains a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| Crotonyl-HIST1H2BC (Lys20) | |
| Polyclonal | |
| Unconjugated | |
| H2BC5 | |
| dJ221C16.3; H2B Clustered Histone 4; H2B histone family, member L; H2B.1; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bc; histone cluster 1, H2bc; histone H2B type 1-C/E/F/G/I; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3017 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.161 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P58876, P62807 | |
| H2BC5, Hist1h2bc | |
| Peptide sequence around site of Crotonyl-Lys (20) derived from Human Histone H2B type 1-C/E/F/G/I. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction