missing translation for 'onlineSavingsMsg'
Learn More

Crotonic Acid, TRC

Product Code. 30041207
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
Unit Size:
10g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041207 10 g 10g
30010151 250 g 250g
30012512 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041207 Supplier Toronto Research Chemicals Supplier No. TRCC81878510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-93-7
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
Synonym 2-Butenoic acid, (E)-, Crotonic acid, (E)- (8CI), (2E)-2-Butenoic acid, (E)-2-Butenoic acid, (E)-Crotonic acid, (E)-Crotonic acid, NSC 8751, trans-2-Butenoic acid, trans-Crotonic acid, (2E)-2-Butenoic acid, (E)-Crotonic acid, trans-2-Butenoic acid, trans-Crotonic acid
IUPAC Name (E)-but-2-enoic acid
SMILES C\C=C\C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H6O2/c1-2-3-4(5)6/h2-3H,1H3,(H,5,6)/b3-2+
Formula Weight 86.0368
Chemical Name or Material Crotonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.