missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotonaldehyde Antibody (2D3.1), Novus Biologicals™
Mouse Monoclonal Antibody
£146.00 - £471.00
Specifications
| Antigen | Crotonaldehyde |
|---|---|
| Clone | 2D3.1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:1000, Dot Blot |
| Applications | Western Blot, ELISA, Dot Blot |
Description
Crotonaldehyde Monoclonal specifically detects Crotonaldehyde in Non-species specific samples. It is validated for Western Blot, ELISA, Dot Blot.Specifications
| Crotonaldehyde | |
| 1 mg/mL | |
| Western Blot, ELISA, Dot Blot | |
| Unconjugated | |
| Mouse | |
| Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH) | |
| Primary | |
| Specific for Crotonaldehyde modified proteins. Does not cross-react with Acrolein, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. |
| 2D3.1 | |
| Western Blot 1:1000, ELISA 1:1000, Dot Blot | |
| Monoclonal | |
| Purified | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title