missing translation for 'onlineSavingsMsg'
Learn More

Crotonaldehyde, 99+%, AcroSeal™, Thermo Scientific Chemicals

Código de producto. 11345546 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Quantity:
100 mL
800 mL
Tamaño de la unidad:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
11345546 100 mL 100mL
10786255 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.
Código de producto. 11345546 Proveedor Thermo Scientific Acros N.º de proveedor 427181000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 4170-30-3
Molecular Weight (g/mol) 70.09
MDL Number MFCD00007003
Synonym crotonaldehyde, 2-butenal, crotylaldehyde, crotonal, 2e-but-2-enal, trans-crotonaldehyde, e-crotonaldehyde, crotonic aldehyde, 1-formylpropene, trans-2-butenal
PubChem CID 447466
ChEBI CHEBI:41607

Especificaciones

Melting Point -74.0°C
Density 0.8580g/mL
Boiling Point 101.0°C to 103.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H6O
Linear Formula CH3CH=CHCHO
Refractive Index 1.4355 to 1.4375
Quantity 100 mL
Merck Index 15, 2587
Specific Gravity 0.858
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,toluene and,kerosene,miscible in all proportions with gasoline,solvent,very soluble in acetone
InChI Key MLUCVPSAIODCQM-NSCUHMNNSA-N
SMILES CC=CC=O
IUPAC Name (E)-but-2-enal
Viscosity 0.27 mPa.s (20°C)
Formula Weight 70.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Crotonaldehyde
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Crotonaldehyde
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H310-Fatal in contact with skin.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.