Learn More
Crotedumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542095
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Crotedumab has been used in trials studying the treatment of Diabetes Mellitus, Type 2.Specifications
Crotedumab | |
Recombinant Monoclonal | |
Unconjugated | |
REGN-1193 | |
Human GCGR | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
2.55 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG4 κ |