missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KVPVHKVGNT PLDMNQFRML FSTCKVPGIT RDSIMNYFRT ESEGRSPNHI VVLCRGRAFV FDVIHEGCLV TPPELLRQLT YIHKKCHSEP DGPGIAALTS EERTRWAKAR EYLIGLD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | CROT |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Crot |
| Gene Accession No. | Q9UKG9 |
| Gene Alias | 1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643 |
| Gene Symbols | Crot |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
