missing translation for 'onlineSavingsMsg'
Learn More

Croscarmellose Sodium, TRC

Product Code. 17929854
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17929854 25 g 25g
17918794 5 g 5g
17918024 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17929854 Supplier Toronto Research Chemicals Supplier No. TRCC81650025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74811-65-7
Synonym Croscarmellose Sodium∣Croscarmellose Sodium

Specifications

Quantity 25 g
Shipping Condition Room Temperature
Chemical Name or Material Croscarmellose Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.