missing translation for 'onlineSavingsMsg'
Learn More

CROP Antibody, Novus Biologicals™

Product Code. 18330087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330087 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330087 Supplier Novus Biologicals Supplier No. NBP303773

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CROP Polyclonal specifically detects CROP in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CROP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, Cisplatin resistance-associated-overexpressed protein, CREAP1, CREAP-1LUC7ACRA, CRE-associated protein 1, CROPFLJ11063, hLuc7A, Luc7A, LUC7-like 3 (S. cerevisiae), luc7-like protein 3, O48, OA48-18, Okadaic acid-inducible phosphoprotein OA48-18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-200 of human CROP (NP_057508.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 51747
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.