missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CROP Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74625AF405
This item is not returnable.
View return policy
Description
CROP Polyclonal antibody specifically detects CROP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CROP | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, Cisplatin resistance-associated-overexpressed protein, CREAP1, CREAP-1LUC7ACRA, CRE-associated protein 1, CROPFLJ11063, hLuc7A, Luc7A, LUC7-like 3 (S. cerevisiae), luc7-like protein 3, O48, OA48-18, Okadaic acid-inducible phosphoprotein OA48-18 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cisplatin resistance-associated overexpressed protein using the numbering given in entry NP_057508.2 (GeneID 51747). | |
| 0.1 mL | |
| Apoptosis | |
| 51747 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction