missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn sodium, MedChemExpress

Product Code. p-200097740
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16450878 5 g 5g
16422139 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16450878

Brand: Medchem Express HYB0320A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cromolyn sodium (Disodium Cromoglycate; FPL-670) is an antiallergic drug. Cromolyn sodium is a GSK-3β inhibitor with an IC50 of 2.0 μM.

Cromolyn sodium (Disodium Cromoglycate

Specifications

Chemical Name or Material Cromolyn sodium
CAS 15826-37-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H14Na2O11
Quantity 5 g
Synonym Disodium Cromoglycate FPL-670
Solubility Information H2O : 50 mg/mL (97.59 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (48.80 mM; Need ultrasonic)
SMILES OC(COC1=C2C(C=C(C(O[Na])=O)OC2=CC=C1)=O)COC3=C4C(C=C(C(O[Na])=O)OC4=CC=C3)=O
Molecular Weight (g/mol) 512.33
Formula Weight 512.33
Percent Purity 98.31%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.