missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn Disodium Salt, TRC

Product Code. 30352737
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352737 50 mg 50mg
30361696 5 g 5g
30356479 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352737 Supplier Toronto Research Chemicals Supplier No. TRCC81500050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15826-37-6
Molecular Formula C23 H14 O11 . 2 Na
Molecular Weight (g/mol) 512.33
Synonym Sodium Cromoglicate, 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid sodium salt (1:2), 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid disodium salt, 5,5'-[(2-Hydroxytrimethylene)dioxy]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid disodium salt, 1,3-Bis(2-carboxychromon-7-yloxy)-2-hydroxypropane disodium salt, 4H-1-Benzopyran-2-carboxylic acid, 5,5'-[(2-hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-, sodium salt (1:2), 4H-1-Benzopyran-2-carboxylic acid, 5,5'-[(2-hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-, disodium salt (9CI), 4H-1-Benzopyran-2-carboxylic acid, 5,5'-[(2-hydroxytrimethylene)dioxy]bis[4-oxo-, disodium salt (8CI), 1,3-Bis(2-carboxychromon-7-yloxy)-2-hydroxypropane disodium salt, Aarane, Alercrom, Alerion, Allergocrom, Altoderm, Colimune, Crolom, Cromoglycate, Cromoglycate disodium, Cromolyn disodium salt, Cromolyn sodium, Cromovet, DNCG, DSCG, Disodium 1,3-bis(2-carboxy-5-chromonyloxy)-2-propanol, Disodium 1,3-bis(2-carboxychromon-5-yloxy)-2-hydroxypropane, Disodium 1,3-bis(2-carboxychromon-5-yloxy)propan-2-ol, Disodium 5,5'-[(2-hydroxytrimethylene)dioxy]bis[4-oxo-4H-1-benzopyran-2-carboxylate], Disodium chromoglycate, Disodium cromoglycate, FPL 670, Fisonair, Fivent, Gastrocrom, Gastrofrenal, Inostral, Intal, Intal Forte, Intal UD, Introl, Irtan, Lomudal, Lomupren, Lomusol, Lomuspray, Nalcrom, Nalcron, NasalCrom, Nasmil, Nasotal, Opticrom, Rynacrom, Sodium cromoglycate, Sodium cromolyn, Sofro, Vividrin
IUPAC Name disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
SMILES [Na+].[Na+].OC(COc1cccc2OC(=CC(=O)c12)C(=O)[O-])COc3cccc4OC(=CC(=O)c34)C(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H16O11.2Na/c24-11(9-31-14-3-1-5-16-20(14)12(25)7-18(33-16)22(27)28)10-32-15-4-2-6-17-21(15)13(26)8-19(34-17)23(29)30;;/h1-8,11,24H,9-10H2,(H,27,28)(H,29,30);;/q;2*+1/p-2
Formula Weight 512.0332
Chemical Name or Material Sodium cromoglicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.