missing translation for 'onlineSavingsMsg'
Learn More

Croconic acid disodium salt, 97%

Product Code. 11485876
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485876 0.25 g 0.25g
11475876 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485876 Supplier Thermo Scientific Alfa Aesar Supplier No. B22235.01
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 14379-00-1 | C5Na2O5 | 186.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14379-00-1
Molecular Formula C5Na2O5
Molecular Weight (g/mol) 186.03
MDL Number MFCD00191954
InChI Key OQXLFPHHAAAVKQ-UHFFFAOYSA-L
Synonym croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt
PubChem CID 12120285
IUPAC Name disodium;3,4,5-trioxocyclopentene-1,2-diolate
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O

Specifications

Melting Point >300°C
Quantity 0.25 g
Formula Weight 186.03
Percent Purity 97%
Chemical Name or Material Croconic acid disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.