missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRNN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11799-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ′fused gene′ family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation.Specifications
| CRNN | |
| Polyclonal | |
| Unconjugated | |
| CRNN | |
| 58 kDa heat shock protein, C1orf10, cornulin, CRNN, DRC1, PDRC1, SEP53, Tumor related protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381457, 49860 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBG3 | |
| Crnn | |
| CRNN Fusion Protein Ag2374 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction