missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRNN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555057
This item is not returnable.
View return policy
Description
Immunogen sequence: YDRQAESQSQ ERISPQIQLS GQTEQTQKAG EGKRNQTTEM RPERQPQTRE QDRAHQTGET VTGSGTQTQA GATQTVEQDS SHQTGRTSKQ TQEATNDQNR Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 40%.
This gene encodes a member of the 'fused gene' family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation.
Specifications
| CRNN | |
| Polyclonal | |
| Unconjugated | |
| CRNN | |
| 53 kDa putative calcium-binding protein; 53 kDa squamous epithelial-induced stress protein; 58 kDa heat shock protein; C1orf10; cornulin; CRNN; DRC1; Gm1015; PDRC1; RGD1311778; SEP53; Squamous epithelial heat shock protein 53; tumor-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 49860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBG3 | |
| CRNN | |
| Recombinant protein corresponding to Human CRNN. Recombinant protein control fragment (Product #RP-89170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction