missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRNN Monoclonal Antibody (OTI4B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527540
This item is not returnable.
View return policy
Description
CRNN Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the 'fused gene' family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation.
Specifications
| CRNN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBG3 | |
| CRNN | |
| Full length human recombinant protein of CRNN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| CRNN | |
| 53 kDa putative calcium-binding protein; 53 kDa squamous epithelial-induced stress protein; 58 kDa heat shock protein; C1orf10; cornulin; CRNN; DRC1; Gm1015; PDRC1; RGD1311778; SEP53; Squamous epithelial heat shock protein 53; tumor-related protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 49860 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu