missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRMP5 Recombinant Rabbit Monoclonal Antibody (23GB5200), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CRMP5 |
|---|---|
| Clone | 23GB5200 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089090
![]() |
Invitrogen™
MA552816 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088747
|
Invitrogen™
MA552817 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
CRMP5 Recombinant Monoclonal antibody specifically detects CRMP5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotEspecificaciones
| CRMP5 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9BPU6, Q9EQF6, Q9JHU0 | |
| 56896, 65208, 65254 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5200 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6 | |
| A synthesized peptide derived from human CRMP5 (500-564AA). | |
| Primary | |
| -20°C | |
| DPYSL5 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto