missing translation for 'onlineSavingsMsg'
Learn More

CRMP4 Antibody, Novus Biologicals™

Product Code. 18381548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381548 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381548 Supplier Novus Biologicals Supplier No. H00001809B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CRMP4 Polyclonal antibody specifically detects CRMP4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_001378.1
Gene Alias Collapsin response mediator protein 4, CRMP-4, CRMP4collapsin response mediator protein 4 long, dihydropyrimidinase-like 3, DRP-3dihydropyrimidinase-related protein 3, DRP3ULIP1, ULIP-1, ULIPLCRMP, Unc-33-like phosphoprotein 1
Host Species Mouse
Immunogen DPYSL3 (NP_001378.1, 1 a.a. - 570 a.a.) full-length human protein. MSYQGKKNIPRITSDRLLIKGGRIVNDDQSFYADIYMEDGLIKQIGDNLIVPGGVKTIEANGKMVIPGGIDVHTHFQMPYKGMTTVDDFFQGTKAALAGGTTMIIDHVVPEPESSLTEAYEKWREWADGKSCCDYALHVDITHWNDSVKQEVQNLIKDKGVNSFMVYMAYKDLYQVSNTELYEIFTCLGELGAIAQVHAENGDIIAQEQTRMLEMGITGPEGHVLSRPEELEAEAVFRAITIASQTNCPLYVTKVMSKSAADLISQARKKGNVVFGEPITASLGIDGTHYWSKNWAKAAAFVTSPPLSPDPTTPDYINSLLASGDLQLSGSAHCTFSTAQKAIGKDNFTAIPEGTNGVEERMSVIWDKAVATGKMDENQFVAVTSTNAAKIFNLYPRKGRISVGSDSDLVIWDPDAVKIVSAKNHQSAAEYNIFEGMELRGAPLVVICQGKIMLEDGNLHVTQGAGRFIPCSPFSDYVYKRIKARRKMADLHAVPRGMYDGPVFDLTTTPKGGTPAGSARGSPTRPNPPVRNLHQSGFSLSGTQVDEGVRSASKRIVAPPGGRSNITSLS
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1809
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.