missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRMP3 Recombinant Rabbit Monoclonal Antibody (23GB1510), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553119
This item is not returnable.
View return policy
Description
Collapsin response mediator proteins (CRMPs), including CRMP-1 (DRP-1), CRMP-2 (DRP-2 or TOAD64), CRMP-3 (DRP-4), CRMP-4 (DRP-3) and CRMP-5 (DRP-5), mediate signal transduction after exposure of neural cells to the axon guidance molecule Semaphorin 3A (SEMA3A)/collapsin. CRMPs are present in the developing cerebral cortex and neocortical neurons and are responsive to SEMA3A. In the adult brain, the expression of CRMPs is dramatically downregulated. However, they remain expressed in structures that retain their capacity for differentiation and plasticity. CRMP-3 is expressed transiently in developing spinal cord and selectively in the postnatal cerebellum.
Specifications
| CRMP3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DPYSL4 | |
| AI173505; Collapsin response mediator protein 3; CRMP3; CRMP-3; dihydropyrimidinase like 4; dihydropyrimidinase-like 4; dihydropyrimidinase-related protein 4; DPY4; Dpys4; DPYSL4; DRP-4; ULIP4; ULIP-4; ULIP4 protein; unc-33-like phosphoprotein 4; UNC33-like phosphoprotein 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10570, 25417, 26757 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1510 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14531, O35098, Q62951 | |
| DPYSL4 | |
| A synthesized peptide derived from human CRMP3 (15-43AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction