missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRMP3 Recombinant Rabbit Monoclonal Antibody (23GB1510), Invitrogen™
GREENER_CHOICE

Product Code. 30087957
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087957 100 μL 100µL
30088792 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087957 Supplier Invitrogen™ Supplier No. MA553118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Collapsin response mediator proteins (CRMPs), including CRMP-1 (DRP-1), CRMP-2 (DRP-2 or TOAD64), CRMP-3 (DRP-4), CRMP-4 (DRP-3) and CRMP-5 (DRP-5), mediate signal transduction after exposure of neural cells to the axon guidance molecule Semaphorin 3A (SEMA3A)/collapsin. CRMPs are present in the developing cerebral cortex and neocortical neurons and are responsive to SEMA3A. In the adult brain, the expression of CRMPs is dramatically downregulated. However, they remain expressed in structures that retain their capacity for differentiation and plasticity. CRMP-3 is expressed transiently in developing spinal cord and selectively in the postnatal cerebellum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1510
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPYSL4
Gene Accession No. O14531, O35098, Q62951
Gene Alias AI173505; Collapsin response mediator protein 3; CRMP3; CRMP-3; dihydropyrimidinase like 4; dihydropyrimidinase-like 4; dihydropyrimidinase-related protein 4; DPY4; Dpys4; DPYSL4; DRP-4; ULIP4; ULIP-4; ULIP4 protein; unc-33-like phosphoprotein 4; UNC33-like phosphoprotein 4
Gene Symbols DPYSL4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CRMP3 (15-43AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10570, 25417, 26757
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.