missing translation for 'onlineSavingsMsg'
Learn More

CRMP2 Rabbit anti-Human, Mouse, Rat, Clone: 0A1T1, Novus Biologicals™

Product Code. 18334302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334302 100 μg 100µL
18362945 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18334302 Supplier Novus Biologicals Supplier No. NBP316406100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CRMP2 Monoclonal antibody specifically detects CRMP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 0A1T1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 473-572 of human CRMP2 (Q16555).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1808
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.