missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP2, p Thr555 Antibody (M539), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23270
This item is not returnable.
View return policy
Description
CRMP2 Monoclonal antibody specifically detects CRMP2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CRMP2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| M539 | |
| Western Blot 1:500 | |
| Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
| Clone M539 was generated from a phospho-CRMP2 (Thr-555) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Thr-555 in human CRMP2. This sequence is conserved in rat and mouse CRMP2 and the phospho-site is not conserved in other CRMP family members. [UniProt# Q16555] | |
| 0.1 mL | |
| Neuroscience, Phospho Specific | |
| 1808 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction