missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRMP2 Monoclonal Antibody (33CT11.7.6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537465
This item is not returnable.
View return policy
Description
CRMP2 Monoclonal Antibody for Western Blot
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
| CRMP2 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O08553, Q16555 | |
| DPYSL2 | |
| KLH conjugated synthetic peptide between 463-492 amino acids from human DRP-2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 33CT11.7.6 | |
| Unconjugated | |
| DPYSL2 | |
| AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406) | |
| Mouse | |
| Protein G | |
| RUO | |
| 12934, 1808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction