missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10317-1-AP-20UL
This item is not returnable.
View return policy
Description
CRMP1 encodes a member of a family of cytosolic phosphoproteins expressed exclusively in the nervous system. The encoded protein is thought to be a part of the semaphorin signal transduction pathway implicated in semaphorin-induced growth cone collapse during neural development. Alternative splicing results in multiple transcript variants.Specifications
| CRMP1 | |
| Polyclonal | |
| Unconjugated | |
| CRMP1 | |
| CRMP 1, CRMP1, DPYSL1, DRP 1, DRP1, ULIP 3, ULIP3, Unc 33 like phosphoprotein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12933, 1400, 25415 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97427, Q14194, Q62950 | |
| Crmp1 | |
| CRMP1 Fusion Protein Ag0369 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction