missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRLF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552453
This item is not returnable.
View return policy
Description
Immunogen sequence: IEHGVNTAED LVREGEIAML GGVGEENEKL WSFTKKASHI QLDSLPEVPL LVDVPCLSAQ LDDSILNIVK DHIFKHGTVA SRPPVQIEEL IEKPGGIIVR WCKVDDDFTA QDYRLQFRKC TSNHFEDVYV GSE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 89%.
May play a role in the negative regulation of cell cycle progression.
Specifications
| CRLF3 | |
| Polyclonal | |
| Unconjugated | |
| CRLF3 | |
| BB164954; Creme9; CREME-9; CRLF3; CRLM9; cytokine receptor like factor 3; cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; cytokine receptor-related factor 4; Cytokine receptor-related protein 4; CYTOR4; FRWS; P48; p48.2; Type I cytokine receptor-like factor p48 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUI8 | |
| CRLF3 | |
| Recombinant protein corresponding to Human CRLF3. Recombinant protein control fragment (Product #RP-89559). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction