missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRLF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103401
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CRLF3.
May play a role in the negative regulation of cell cycle progression.
Specifications
| CRLF3 | |
| Polyclonal | |
| Unconjugated | |
| CRLF3 | |
| BB164954; Creme9; CREME-9; CRLF3; CRLM9; cytokine receptor like factor 3; cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; cytokine receptor-related factor 4; Cytokine receptor-related protein 4; CYTOR4; FRWS; P48; p48.2; Type I cytokine receptor-like factor p48 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51379, 54394, 54395 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IUI8, Q9Z2L7 | |
| CRLF3 | |
| A synthesized peptide derived from human CRLF3(Accession Q8IUI8), corresponding to amino acid residues G302-V352. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction