missing translation for 'onlineSavingsMsg'
Learn More

CRLF1 Antibody (4F4), Novus Biologicals™

Product Code. 18379478 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379478 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379478 Supplier Novus Biologicals Supplier No. H00009244M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRLF1 Monoclonal antibody specifically detects CRLF1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRLF1
Applications Western Blot, ELISA, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 4F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004741
Gene Alias CISS1, CISScytokine type 1 receptor CRLP-1, CLF, CLF-1class I cytokine receptor, cytokine receptor-like factor 1, Cytokine-like factor 1, NR6, ZcytoR5
Host Species Mouse
Immunogen CRLF1 (NP_004741, 135 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PEKPVNISCWSKNMKDLTCRWTPGAHGETFLHTNYSLKYKLRWYGQDNTCEEYHTVGPHSCHIPKDLALFTPYEIWVEATNRLGSARSDVLTLDIL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9244
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.