missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CrkRS Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-87011H
This item is not returnable.
View return policy
Description
CrkRS Polyclonal antibody specifically detects CrkRS in Human samples. It is validated for Western Blot
Specifications
| CrkRS | |
| Polyclonal | |
| Western Blot | |
| Cdc2-related kinase, arginine/serine-richCrkRS, Cell division cycle 2-related protein kinase 7, Cell division protein kinase 12, CRK7CDC2-related protein kinase 7, CRKR, CRKRS, cyclin-dependent kinase 12, EC 2.7.11, EC 2.7.11.23, hCDK12, KIAA0904EC 2.7.11.22 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cell division cycle 2 (CDC2)-related kinase arginine/serine-rich using the numbering given in entry NP_057591.2 (GeneID 51755). | |
| 0.1 mL | |
| Protein Kinase | |
| 51755 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction