missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crk, p Tyr221 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23252
This item is not returnable.
View return policy
Description
Crk Polyclonal antibody specifically detects Crk in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot
Specifications
| Crk | |
| Polyclonal | |
| Western Blot 1:1000 | |
| adapter molecule crk, avian sarcoma virus CT10 (v-crk) oncogene homolog, CRKII, p38, Proto-oncogene c-Crk, v-crk avian sarcoma virus CT10 oncogene homolog, v-crk sarcoma virus CT10 oncogene homolog (avian) | |
| Phospho-Crk (Tyr-221) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Tyr-221 in human Crk II. This peptide sequence is well conserved in mouse and rat Crk II, and has 40% homology with the conserved site in CrkL (Tyr-207). The site is not found in Crk I. [UniProt# P46108] | |
| 0.1 mL | |
| Apoptosis, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| 1398 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction