missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cpf1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62735DL594
This item is not returnable.
View return policy
Description
CRISPR-Cpf1 Polyclonal specifically detects CRISPR-Cpf1 in Bacterial samples. It is validated for Western Blot.
Specifications
| CRISPR-Cpf1 | |
| Polyclonal | |
| Western Blot | |
| Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
| Partial recombinant Francisella tularensis protein (between amino acids 600-850) [NCBI WP_003034647.1] | |
| 0.1 mL | |
| Bacteria | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction