missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cpf1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62735
311.95 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CRISPR-Cpf1 Polyclonal antibody specifically detects CRISPR-Cpf1 in Bacteria samples. It is validated for Western Blot
Specifications
| CRISPR-Cpf1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| anti-cas12a, anti-CPF1, anti-CRISPR-Cas12a, anti-CRISPR-Cpf1, Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
| Partial recombinant Francisella tularensis protein (between amino acids 600-850) [NCBI WP_003034647.1] | |
| 0.1 mg | |
| Cell Biology, DNA Repair, Virology Bacteria and Parasites | |
| Bacteria | |
| Purified |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1.0 μg/mL | |
| A0Q7Q2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction