missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cpf1 Antibody (3D3-F7), Janelia Fluor™ 646 , Novus Biologicals™

Product Code. 18413625 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413625 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413625 Supplier Novus Biologicals Supplier No. NBP278820JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRISPR-Cpf1 Monoclonal antibody specifically detects CRISPR-Cpf1 in Bacteria samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISPR-Cpf1
Applications Western Blot
Classification Monoclonal
Clone 3D3-F7
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1
Host Species Mouse
Immunogen Partial recombinant protein made to a C-terminal portion of the Acidaminococcus sp. CRISPR-Cpf1 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, DNA Repair, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.