missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cpf1 Antibody (2D5-6G11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-78819AF700
This item is not returnable.
View return policy
Description
CRISPR-Cpf1 Monoclonal specifically detects CRISPR-Cpf1 in Bacteria, Bacterial samples. It is validated for Western Blot.
Specifications
| CRISPR-Cpf1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This antibody is specific for Lachnospiraceae bacterium CRISPR-Cpf1. | |
| Store at 4°C in the dark. |
| Western Blot | |
| 2D5-6G11 | |
| Western Blot | |
| Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
| Partial recombinant protein made to an N-terminal portion of the Lachnospiraceae bacterium CRISPR-Cpf1 protein | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction