missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18421019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421019 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421019 Supplier Novus Biologicals Supplier No. NBP305548AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISPR-Cas9
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Host Species Rabbit
Immunogen N-terminal region, amino acids 1-608 and C-terminal region, amino acids 814-1372 of S. pyogenes CRISPR-CAS9 sequence CDJ55032.1 from S. pyogenes, expressed in and purified from E. coli
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Gene ID (Entrez) 901176
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.