missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody (7A9-3A3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-36440F

Product Code. 18648805

  • £520.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Mouse, Bacteria, Bovine, Chicken, Fungi samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry, KnockDown
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRISPR-Cas9
Monoclonal
FITC
PBS
Mouse
Protein G purified
RUO
Primary
Mouse, Bacteria, Bovine, Chicken, Fungi, Chicken, Equine, Primate, Xenopus species, Zebrafish
Purified
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry, KnockDown
7A9-3A3
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount, Knockout Validated
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2].
0.1 mL
Gene Editing, Gene Knockouts, Gene Therapy
901176
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.