missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody (6H4), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18330341 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330341 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330341 Supplier Novus Biologicals Supplier No. NBP281124AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISPR-Cas9
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 6H4
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Host Species Mouse
Immunogen Partial recombinant protein made to an N terminal portion of the Staphylococcus aureus CRISPR/Cas9 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Gene ID (Entrez) 901176
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.