Learn More
Invitrogen™ CRISPLD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114202
Description
Immunogen sequence: YSPKGNWWGH APYKHGRPCS ACPPSFGGGC RENLCYKEGS DRYYPPREEE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Yeast: 82%.
Specifications
| CRISPLD1 | |
| Polyclonal | |
| Unconjugated | |
| CRISPLD1 | |
| CocoaCrisp; CRISP10; CRISP-10; CRISPLD1; cysteine rich secretory protein LCCL domain containing 1; cysteine-rich secretory protein 10; cysteine-rich secretory protein LCCL domain containing 1; cysteine-rich secretory protein LCCL domain-containing 1; LCCL domain containing cysteine-rich secretory protein 1; LCCL domain-containing cysteine-rich secretory protein 1; LCRISP1; RGD1564813; Trypsin inhibitor Hl; UNQ342/PRO541 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83690 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H336 | |
| CRISPLD1 | |
| Immunogen sequence: YSPKGNWWGH APYKHGRPCS ACPPSFGGGC RENLCYKEGS DRYYPPREEE (aa 206-255). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.