missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRISP2 Polyclonal Antibody

Product Code. 12859152
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859152 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859152 Supplier Invitrogen™ Supplier No. PA521007

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human testis tissue lysate. PA5-21007 can be used with blocking peptide PEP-1121.

The cysteine-rich secretory proteins (CRISP) family is a group of four proteins that are strongly expressed in the male reproductive tract and have been implicated in having roles in male fertility. CRISP2, also known as TPX1, has been implicated in the adhesion between spermatids and Sertoli cells, and with CRISP1, is thought to be involved in sperm-egg fusion. CRISP2 has been shown to regulate the Ca2+ influx through ryanodine receptors (RYR) and may influence the acrosome reaction or sperm motility. CRISP2 has also been shown to bind to the mitogen-activated protein kinase kinase kinase 11 (MAP3K11) and localizes to the developing acrosome, suggesting this CRISP2-MAP3K11 complex may have a role in acrosome development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISP2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CRISP2
Gene Accession No. P16562
Gene Alias Cancer/testis antigen 36; CRISP2; CRISP-2; Crisp2l; CT36; cysteine rich secretory protein 2; cysteine-rich secretory protein 2; cysteine-rich secretory protein 2-like; GAPDL5; glyceraldehyde-3-phosphate dehydrogenase-like 5; MGC111136; OTTHUMP00000016589; testicular tissue protein Li 43; testis specific gene 1; testis specific protein 1; testis specific protein 1 (probe H4-1 p3-1); testis-specific protein TPX-1; TPX1; Tpx-1; TSP1
Gene Symbols CRISP2
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of human CRISP2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7180
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.