missing translation for 'onlineSavingsMsg'
Learn More

CRISP-3 Antibody (295220) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30735408 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735408 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735408 Supplier Novus Biologicals Supplier No. FAB2397MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRISP-3 Monoclonal antibody specifically detects CRISP-3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRISP-3
Applications Western Blot
Classification Monoclonal
Clone 295220
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRISP-3, Asn21-Tyr245 (Ser106Pro & Ala134Ser), Accession # P54108.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10321
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.