missing translation for 'onlineSavingsMsg'
Learn More

Crisaborole, TRC

Product Code. 30395965
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395965 250 mg
30389228 25 mg
30408158 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395965 Supplier Toronto Research Chemicals Supplier No. TRCH947928250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 906673-24-3
Molecular Formula C14H10BNO3
Molecular Weight (g/mol) 251.05
Synonym ,4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)benzonitrile, 4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]benzonitrile, 5-(4-Cyanophenoxy)-1,3-dihydro-1-hydroxy-[2,1]-benzoxaborole, 5-(4-Cyanophenoxy)-2,3-dihydro-1-hydroxy-2,1-benzoxaborole, AN2728
IUPAC Name 4-[(1-hydroxy-3H-2,1-benzoxaborol-5-yl)oxy]benzonitrile
SMILES OB1OCc2cc(Oc3ccc(cc3)C#N)ccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H10BNO3/c16-8-10-1-3-12(4-2-10)19-13-5-6-14-11(7-13)9-18-15(14)17/h1-7,17H,9H2
Formula Weight 251.0754
Chemical Name or Material Crisaborole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.