missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIPTO Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1598H
This item is not returnable.
View return policy
Description
CRIPTO Polyclonal specifically detects CRIPTO in Human, Mouse samples. It is validated for Western Blot.
Specifications
| CRIPTO | |
| Polyclonal | |
| Western Blot | |
| P13385 | |
| TDGF1 | |
| A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50). [UniProt# P51865] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1 | |
| Rabbit | |
| 18 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 6997 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction