missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIPTO Antibody (89633) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2772JF525
This item is not returnable.
View return policy
Description
CRIPTO Monoclonal antibody specifically detects CRIPTO in Human,Primate samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),Immunocytochemistry,CyTOF
Specifications
| CRIPTO | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1 | |
| E. coli-derived recombinant human Cripto-1, Arg38-Tyr188, Accession # P13385 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Neuroscience, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 6997 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF | |
| 89633 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction