missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRIPT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566039
This item is not returnable.
View return policy
Description
Immunogen sequence: CAYKKGICAMC GKKVLDTKNY KQTSV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.
Specifications
| CRIPT | |
| Polyclonal | |
| Unconjugated | |
| CRIPT | |
| 1200020A08Rik; AA589469; CRIPT; CXXC repeat containing interactor of PDZ3 domain; cysteine-rich interactor of PDZ three; Cysteine-rich interactor of PDZ3; cysteine-rich PDZ-binding protein; HSPC139; postsynaptic protein CRIPT; SSMDF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9419 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P021 | |
| CRIPT | |
| Recombinant Human CRIPT. Recombinant protein control fragment (Product #RP-106730). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction