missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRIP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559984

Product Code. 15747831

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PCYAAMFGPK GFGRGGAESH TFK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

CRIP1 is a cysteine-rich intestinal protein (CRIP) that belongs to the LIM/double zinc finger protein family, members of which include cysteine- and glycine-rich protein-1 (CSRP1 MIM 123876), rhombotin-1 (RBTN1 MIM 186921), rhombotin-2 (RBTN2 MIM 180385), and rhombotin-3 (RBTN3 MIM 180386). CRIP may be involved in intestinal zinc transport.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRIP1
Polyclonal
Unconjugated
CRIP1
CRHP; Crip; CRIP1; CRP1; CRP-1; cysteine rich intestinal protein; cysteine rich protein 1; cysteine-rich heart protein; Cysteine-rich intestinal protein; cysteine-rich protein 1; cysteine-rich protein 1 (intestinal)
Rabbit
Antigen affinity chromatography
RUO
1396
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P50238
CRIP1
Recombinant protein corresponding to Human CRIP1. Recombinant protein control fragment (Product #RP-99286).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.