Learn More
JG Finneran Associates™ Crimp Caps
Product Code. 10701242
Click to view available options
Color:
Clear
Red
Red, Blue
Red, Clear
Red, Green
Red, Silver
Red, Yellow
Silver
Closure Size:
11 mm
20 mm
Material:
PTFE, Aluminum
PTFE, Rubber, Aluminum
PTFE, Rubber, Poly
PTFE, Silicone, Aluminum
PTFE, Silicone, Poly
Unit Size:
Pack of 1000
Product Code. 10701242
Brand: JG Finneran Associates™ 511020
Only null left
Add to basket
Add to basket
Use these caps with standard 11mm chromatography vials. JG Finneran™ Crimp Caps feature an open top for filling and retrieving samples with a syringe.
  • Made of the highest quality aluminum
  • With PTFE seal
  • Seals are available in a variety of colors for easy sample identification
Specifications
Material (Liner) PTFE
Color Silver
Material PTFE, Aluminum
Septum PTFE seal
Quantity 1000 Pk.
Type Crimp Cap
Closure Size 20 mm