missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Crimp Caps

Use these caps with standard 11mm chromatography vials. JG Finneran™ Crimp Caps feature an open top for filling and retrieving samples with a syringe.

Brand:  JG Finneran Associates™ 5140-11

Code : 17

Additional Details : Weight : 0.24300kg

 View more versions of this product

Product Code. 10401021

  • £451.00 / Pack of 1000
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Made of the highest quality aluminum
  • With PTFE seal
  • Seals are available in a variety of colors for easy sample identification
Specifications

Specifications

Rubber
PTFE, Rubber, Aluminum
1000 Pk.
11 mm
Red, Silver
Clear PTFE/red Rubber
Crimp Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers