missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesis™ Crimp Cap, 11 mm, White Silicone/White PTFE Septa
Click to view available options
Color:
White
Unit Size:
Pack of 1000
Description
- White silicone cap
- White PTFE septa
- Secure closure
- Suitable for autosampler vials
Specifications
Specifications
| Color | White |
| Material | Silicone, PTFE, Aluminum |
| For Use With (Application) | Capping vials, providing septa access |
| For Use With (Equipment) | Vials with Neck Diameter of 11 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction