missing translation for 'onlineSavingsMsg'
Learn More

CRIM1 Antibody (MM0202-16G1), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18426106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426106 Supplier Novus Biologicals Supplier No. NBP212207JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRIM1 Monoclonal antibody specifically detects CRIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIM1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0202-16G1
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52
Host Species Mouse
Immunogen Human recombinant CRIM1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51232
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.