missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRIF1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
CRIF1 Polyclonal antibody specifically detects CRIF1 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | CRIF1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | CKBBP2, CKII beta-associating protein, CR6 interacting factor 1, CRIF1p53-responsive gene 6 protein, growth arrest and DNA damage-inducible proteins-interacting protein 1, growth arrest and DNA-damage-inducible, gamma interacting protein 1, MGC4667, MGC4758, papillomavirus L2 interacting nuclear protein 1, Papillomavirus L2-interacting nuclear protein 1, Plinp1, PLINP-1CKII beta binding protein 2, PRG6CR6-interacting factor 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: WPDPEDLLTPRWQLGPRYAAKQFARYGAASGVVPGSLWPSPE |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?